Assay Details
Gene Symbol: TSPAN16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682378.2 | XP_005682435.1 | 4 - 5 | 532 | 83 | |
XM_018051744.1 | XP_017907233.1 | 5 - 6 | 607 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
