Assay Details
| Gene Symbol: BEND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684604.3 | XP_005684661.2 | 3 - 4 | 653 | 65 | |
| XM_018053103.1 | XP_017908592.1 | 3 - 4 | 530 | 65 | ||
More Information
Set Membership: |
|

BEND3
Genomic Map