Assay Details
| Gene Symbol: MRPL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684948.3 | XP_005685005.1 | 1 - 2 | 178 | 65 | |
| XM_018053433.1 | XP_017908922.1 | 1 - 2 | 185 | 65 | ||
More Information
Set Membership: |
|

MRPL18
Genomic Map