Assay Details
Gene Symbol: VCPKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685632.3 | XP_005685689.2 | 4 - 5 | 1080 | 98 | |
XM_013967055.2 | XP_013822509.2 | 3 - 4 | 998 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
