Assay Details
Gene Symbol: CMTM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062088.1 | XP_017917577.1 | 4 - 5 | 647 | 61 | |
GenBank mRNA | JO392005.2 | - | 1 - 2 | 400 | 61 | |
JO582121.1 | - | 4 - 5 | 756 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
