Assay Details
| Gene Symbol: ADGRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696383.3 | XP_005696440.2 | 2 - 3 | 1150 | 71 | |
| XM_005696384.3 | XP_005696441.2 | 2 - 3 | 318 | 71 | ||
| XM_005696385.3 | XP_005696442.2 | 2 - 3 | 156 | 71 | ||
| GenBank mRNA | KA352595.1 | - | 2 - 3 | 316 | 71 | |
More Information
Set Membership: |
|

ADGRF2
Genomic Map