Assay Details
Gene Symbol: ADGRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696383.3 | XP_005696440.2 | 4 - 5 | 1344 | 73 | |
XM_005696384.3 | XP_005696441.2 | 4 - 5 | 512 | 73 | ||
XM_005696385.3 | XP_005696442.2 | 4 - 5 | 350 | 73 | ||
XM_005696386.3 | XP_005696443.2 | 4 - 5 | 471 | 73 | ||
XM_005696387.3 | XP_005696444.2 | 2 - 3 | 209 | 73 | ||
XM_018039214.1 | XP_017894703.1 | 3 - 4 | 293 | 73 | ||
GenBank mRNA | KA352595.1 | - | 4 - 5 | 510 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
