Assay Details
Gene Symbol: TSPAN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048625.1 | XP_017904114.1 | 1 - 2 | 929 | 70 | |
XM_018048626.1 | XP_017904115.1 | 3 - 4 | 229 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
