Assay Details
| Gene Symbol: LOC102187310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO633254.2 | - | 314 | 57 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102187310| Gene Symbol: LOC102187310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO633254.2 | - | 314 | 57 | ||