Assay Details
Gene Symbol: ARSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053677.1 | XP_017909166.1 | 4 - 5 | 1044 | 63 | |
GenBank mRNA | JO599198.2 | - | 3 - 4 | 613 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
