Assay Details
| Gene Symbol: NACC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056079.1 | XP_017911568.1 | 3 - 4 | 1223 | 85 | |
| XM_018056081.1 | XP_017911570.1 | 3 - 4 | 1626 | 85 | ||
More Information
Set Membership: |
|

NACC2
Genomic Map