Assay Details
| Gene Symbol: ACBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060225.1 | XP_017915714.1 | 4 - 5 | 846 | 65 | |
| GenBank mRNA | JO365657.2 | - | 4 - 5 | 1428 | 65 | |
| JO500843.2 | - | 706 | 65 | |||
More Information
Set Membership: |
|

ACBD3
Genomic Map