Assay Details
| Gene Symbol: SGF29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040164.1 | XP_017895653.1 | 9 - 10 | 961 | 78 | |
| XM_018040165.1 | XP_017895654.1 | 10 - 11 | 1001 | 78 | ||
| GenBank mRNA | JO628882.2 | - | 1 - 2 | 176 | 78 | |
More Information
Set Membership: |
|

SGF29
Genomic Map