Assay Details
Gene Symbol: CYTIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676115.3 | XP_005676172.1 | 6 - 7 | 622 | 76 | |
GenBank mRNA | JT677555.1 | - | 6 - 7 | 461 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
