Assay Details
Gene Symbol: LGALS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681055.3 | XP_005681112.1 | 2 - 3 | 157 | 68 | |
XM_013964330.2 | XP_013819784.1 | 2 - 3 | 556 | 68 | ||
GenBank mRNA | JO422482.2 | - | 2 - 3 | 469 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
