Assay Details
| Gene Symbol: DMRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683769.3 | XP_005683826.2 | 3 - 4 | 815 | 61 | |
| GenBank mRNA | JT715356.1 | - | 1 - 2 | 155 | 61 | |
More Information
Set Membership: |
|

DMRT1
Genomic Map