Assay Details
| Gene Symbol: CHMP4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057730.1 | XP_017913219.1 | 4 - 5 | 771 | 73 | |
| GenBank mRNA | JO579103.1 | - | 1 - 2 | 328 | 73 | |
| KA324300.1 | - | 4 - 5 | 836 | 73 | ||
More Information
Set Membership: |
|

CHMP4B
Genomic Map