Assay Details
Gene Symbol: TMEM171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005694562.3 | XP_005694619.2 | 2 - 3 | 2957 | 72 | |
XM_018065685.1 | XP_017921174.1 | 3 - 4 | 1410 | 72 | ||
XM_018065686.1 | XP_017921175.1 | 3 - 4 | 1170 | 72 | ||
XM_018065688.1 | XP_017921177.1 | 3 - 4 | 994 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
