Assay Details
Gene Symbol: DDIAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699482.3 | XP_005699539.2 | 5 - 6 | 627 | 90 | |
XM_018043429.1 | XP_017898918.1 | 3 - 4 | 581 | 90 | ||
XM_018043430.1 | XP_017898919.1 | 4 - 5 | 498 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
