Assay Details
| Gene Symbol: NDUFB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046860.1 | XP_017902349.1 | 2 - 3 | 321 | 61 | |
| GenBank mRNA | JO426456.1 | - | 2 - 3 | 363 | 61 | |
| JO505411.2 | - | 2 - 3 | 287 | 61 | ||
More Information
Set Membership: |
|

NDUFB2
Genomic Map