Assay Details
Gene Symbol: GZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057276.1 | XP_017912765.1 | 2 - 3 | 1472 | 70 | |
XM_018057277.1 | XP_017912766.1 | 2 - 3 | 1820 | 70 | ||
XM_018057278.1 | XP_017912767.1 | 2 - 3 | 1563 | 70 | ||
XM_018057279.1 | XP_017912768.1 | 2 - 3 | 1462 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
