Assay Details
| Gene Symbol: RNF114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058014.1 | XP_017913503.1 | 3 - 4 | 449 | 64 | |
| GenBank mRNA | JO623737.2 | - | 2 - 3 | 301 | 64 | |
| JO623738.2 | - | 2 - 3 | 301 | 64 | ||
More Information
Set Membership: |
|

RNF114
Genomic Map