Assay Details
Gene Symbol: LOC102174762 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058179.1 | XP_017913668.1 | 3 - 4 | 516 | 73 | |
GenBank mRNA | JO612734.2 | - | 5 - 6 | 886 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
