Assay Details
Gene Symbol: RPP40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039263.1 | XP_017894752.1 | 6 - 7 | 797 | 73 | |
GenBank mRNA | KA339571.1 | - | 2 - 3 | 244 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
