Assay Details
| Gene Symbol: SLC25A16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013975663.2 | XP_013831117.2 | 7 - 8 | 913 | 78 | |
| XM_018042584.1 | XP_017898073.1 | 6 - 7 | 532 | 78 | ||
| XM_018042585.1 | XP_017898074.1 | 7 - 8 | 913 | 78 | ||
More Information
Set Membership: |
|

SLC25A16
Genomic Map