Assay Details
| Gene Symbol: STMN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057526.1 | XP_017913015.1 | 2 - 3 | 325 | 60 | |
| GenBank mRNA | JO635272.2 | - | 3524 | 60 | ||
More Information
Set Membership: |
|

STMN3
Genomic Map