Assay Details
Gene Symbol: GPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061975.1 | XP_017917464.1 | 5 - 6 | 730 | 62 | |
GenBank mRNA | KA322812.1 | - | 5 - 6 | 776 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
