Assay Details
Gene Symbol: SMIM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695872.3 | XP_005695929.1 | 1 - 2 | 393 | 102 | |
XM_005695873.3 | XP_005695930.2 | 1 - 2 | 393 | 102 | ||
GenBank mRNA | JO458543.2 | - | 1 - 2 | 345 | 102 | |
JT727572.1 | - | 2 - 3 | 316 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
