Assay Details
Gene Symbol: KAZALD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698401.3 | XP_005698458.2 | 4 - 5 | 1022 | 64 | |
GenBank mRNA | JO581711.1 | - | 1 - 2 | 1660 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
