Assay Details
Gene Symbol: AKAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053358.1 | XP_017908847.1 | 4 - 5 | 5483 | 65 | |
XM_018053359.1 | XP_017908848.1 | 2 - 3 | 5209 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
