Assay Details
| Gene Symbol: ST3GAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061683.1 | XP_017917172.1 | 3 - 4 | 1694 | 62 | |
| XM_018061684.1 | XP_017917173.1 | 4 - 5 | 1856 | 62 | ||
| XM_018061685.1 | XP_017917174.1 | 3 - 4 | 1678 | 62 | ||
| GenBank mRNA | JO329394.2 | - | 4 - 5 | 782 | 62 | |
| JO424989.1 | - | 3 - 4 | 1796 | 62 | ||
More Information
Set Membership: |
|

ST3GAL2
Genomic Map