Assay Details
| Gene Symbol: RSPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675639.3 | XP_005675696.1 | 4 - 5 | 580 | 66 | |
| XM_005675640.3 | XP_005675697.1 | 3 - 4 | 466 | 66 | ||
| GenBank mRNA | JO562478.2 | - | 5 - 6 | 681 | 66 | |
More Information
Set Membership: |
|

RSPH1
Genomic Map