Assay Details
| Gene Symbol: C2H2orf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066588.1 | XP_017922077.1 | 2 - 3 | 192 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2H2orf66| Gene Symbol: C2H2orf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066588.1 | XP_017922077.1 | 2 - 3 | 192 | 84 | |
Set Membership: |
|