Assay Details
| Gene Symbol: SDHC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677156.3 | XP_005677213.1 | 4 - 5 | 299 | 57 | |
| GenBank mRNA | JO581307.1 | - | 2 - 3 | 1120 | 57 | |
More Information
Set Membership: |
|

SDHC
Genomic Map