Assay Details
Gene Symbol: RIPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685203.3 | XP_005685260.2 | 7 - 8 | 1195 | 61 | |
XM_013966824.2 | XP_013822278.2 | 7 - 8 | 1192 | 61 | ||
XM_013966825.2 | XP_013822279.2 | 6 - 7 | 1053 | 61 | ||
XM_018054645.1 | XP_017910134.1 | 7 - 8 | 1210 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
