Assay Details
| Gene Symbol: BATF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686099.3 | XP_005686156.1 | 1 - 2 | 306 | 80 | |
| GenBank mRNA | JO612918.2 | - | 1 - 2 | 309 | 80 | |
More Information
Set Membership: |
|

BATF
Genomic Map