Assay Details
Gene Symbol: LGALS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692379.3 | XP_005692436.1 | 9 - 10 | 1227 | 63 | |
XM_018062361.1 | XP_017917850.1 | 10 - 11 | 1148 | 63 | ||
XM_018062363.1 | XP_017917852.1 | 10 - 11 | 1086 | 63 | ||
GenBank mRNA | KA305428.1 | - | 9 - 10 | 1215 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
