Assay Details
Gene Symbol: XAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693424.3 | XP_005693481.2 | 4 - 5 | 625 | 59 | |
XM_005693425.3 | XP_005693482.2 | 4 - 5 | 625 | 59 | ||
GenBank mRNA | JO423365.2 | - | 5 - 6 | 850 | 59 | |
JO587107.2 | - | 4 - 5 | 460 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
