Assay Details
| Gene Symbol: FANCD2OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066744.1 | XP_017922233.1 | 2 | 479 | 85 | |
| XM_018066745.1 | XP_017922234.1 | 2 | 634 | 85 | ||
More Information
Set Membership: |
|

FANCD2OS
Genomic Map