Assay Details
| Gene Symbol: LOC102187823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045840.1 | XP_017901329.1 | 5 - 6 | 2040 | 86 | |
| XM_018045841.1 | XP_017901330.1 | 4 - 5 | 1884 | 86 | ||
More Information
Set Membership: |
|

LOC102187823
Genomic Map