Assay Details
| Gene Symbol: TNIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049837.1 | XP_017905326.1 | 3 - 4 | 778 | 55 | |
| GenBank mRNA | JO630007.2 | - | 3 - 4 | 776 | 55 | |
| KA331611.1 | - | 3 - 4 | 1088 | 55 | ||
More Information
Set Membership: |
|

TNIP2
Genomic Map