Assay Details
| Gene Symbol: LTC4S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051604.1 | XP_017907093.1 | 1 - 2 | 157 | 74 | |
| GenBank mRNA | KA336110.1 | - | 1 - 2 | 171 | 74 | |
More Information
Set Membership: |
|

LTC4S
Genomic Map