Assay Details
| Gene Symbol: WNT9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051577.1 | XP_017907066.1 | 3 - 4 | 698 | 82 | |
| GenBank mRNA | JO642354.2 | - | 2 - 3 | 367 | 82 | |
More Information
Set Membership: |
|

WNT9A
Genomic Map