Assay Details
Gene Symbol: TRIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052215.1 | XP_017907704.1 | 4 - 5 | 825 | 64 | |
XM_018052216.1 | XP_017907705.1 | 3 - 4 | 729 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
