Assay Details
| Gene Symbol: SLC35D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052476.1 | XP_017907965.1 | 7 - 8 | 641 | 76 | |
| XM_018052477.1 | XP_017907966.1 | 6 - 7 | 572 | 76 | ||
| XM_018052478.1 | XP_017907967.1 | 7 - 8 | 642 | 76 | ||
| XM_018052479.1 | XP_017907968.1 | 5 - 6 | 500 | 76 | ||
More Information
Set Membership: |
|

SLC35D2
Genomic Map