Assay Details
| Gene Symbol: FCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056197.1 | XP_017911686.1 | 1 - 2 | 165 | 129 | |
| GenBank mRNA | JO340138.2 | - | 8 - 9 | 1079 | 129 | |
More Information
Set Membership: |
|

FCN1
Genomic Map