Assay Details
| Gene Symbol: FCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056197.1 | XP_017911686.1 | 7 - 8 | 667 | 110 | |
| GenBank mRNA | JO340138.2 | - | 2 - 3 | 577 | 110 | |
| KA353194.1 | - | 4 - 5 | 324 | 110 | ||
More Information
Set Membership: |
|

FCN1
Genomic Map