Assay Details
Gene Symbol: BCL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061300.1 | XP_017916789.1 | 2 - 3 | 324 | 88 | |
XM_018061301.1 | XP_017916790.1 | 2 - 3 | 322 | 88 | ||
GenBank mRNA | KA335294.1 | - | 4 - 5 | 1756 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
