Assay Details
| Gene Symbol: BCL7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061300.1 | XP_017916789.1 | 3 - 4 | 422 | 66 | |
| XM_018061301.1 | XP_017916790.1 | 3 - 4 | 420 | 66 | ||
| GenBank mRNA | KA335294.1 | - | 3 - 4 | 1658 | 66 | |
More Information
Set Membership: |
|

BCL7A
Genomic Map