Assay Details
| Gene Symbol: COG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062208.1 | XP_017917697.1 | 3 - 4 | 1482 | 64 | |
| GenBank mRNA | JO621286.2 | - | 2 - 3 | 622 | 64 | |
More Information
Set Membership: |
|

COG8
Genomic Map